Learn More
Fisherbrand™ Polystyrene Storage Bottles
GREENER_CHOICE
Catalog No. FB12566516
Change view
Click to view available options
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
FB12566516 1000 mL
FB12566513 150 mL
FB12566514 250 mL
FB12566515 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FB12566516 Supplier Fisherbrand™ Supplier No. FB12566516
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Individually packaged sterile screw top style bottle cap
Reduce the risk of cross contamination with sterile, disposable polystyrene media bottles. Use with Bottle-Top Filters or as replacement/extra receptacles for Filter Units. Excellent as single-use storage containers.
  • Rounded with molded in hand grips
  • Polystyrene
  • Molded in graduations
  • Sterile
Certifications
Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility
Specifications
Description Rounded with molded in hand grips
Material Polystyrene
Sterility Sterile
Type Receiver Flask
Volume (Metric) 1000 mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.