missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Polystyrene Sterile Single Wrapped Reagent Reservoir

Catalog No. 03449266
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
55 mL
100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03449266 100 mL
03449255 5 mL
03449257 10 mL
03449259 25 mL
03449262 50 mL
03449278 55 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03449266 Supplier Heathrow Scientific™ Supplier No. 120648

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

  • Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile, sealed in bags and electron beam sterilized
  • DNase, RNase and Pyrogen free
  • Sealed in easy-to-open bags
  • Quality certificate available on request
  • Disposable

Specifications

Capacity (English) 3.4 oz.
Color White
Capacity (Metric) 100 mL
Dimensions (L x W x H) 4.3 x 1.9 x 1.3 in.
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Packaging Single Wrapped
Quantity 1/Bag, 100 Bags/Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Convoluted
Show More Show Less