Learn More
Cole-Parmer™ Essentials Pipette Basins
V-shaped design minimizes reagent waste
$102.13 - $252.66
Specifications
| Color | White |
|---|---|
| Material | Polystyrene |
| Type | Pipetting Reservoir |
| For Use With (Application) | Holding Reagent |
| Autoclavable | Not Autoclavable |
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | No. per Case | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | No. per Case | Price | Quantity | |||
|
03-391-535
|
Cole-Parmer™ Essentials
0439523 |
25 mL | Sterile | 50 |
Case of 50 Each for $102.13
|
|
|||
|
03-391-536
|
Cole-Parmer™ Essentials
0439524 |
25 mL | Sterile | 100 |
Case of 100 Each for $207.70
|
|
|||
|
03-391-537
|
Cole-Parmer™ Essentials
0439525 |
25 mL | Non-sterile | 100 |
Case of 100 Each for $165.91
|
|
|||
|
03-391-539
|
Cole-Parmer™ Essentials
0439527 |
50 mL | Sterile | 100 |
Case of 100 Each for $219.98
|
|
|||
|
03-391-538
|
Cole-Parmer™ Essentials
0439526 |
50 mL | Sterile | 50 |
Case of 50 Each for $108.19
|
|
|||
|
03-391-540
|
Cole-Parmer™ Essentials
0439528 |
50 mL | Non-sterile | 100 |
Case of 100 Each for $187.42
|
|
|||
|
03-391-541
|
Cole-Parmer™ Essentials
0439529 |
100 mL | Sterile | 50 |
Case of 50 Each for $117.66
|
|
|||
|
03-391-542
|
Cole-Parmer™ Essentials
0439530 |
100 mL | Sterile | 100 |
Case of 100 Each for $252.66
|
|
|||
|
03-391-543
|
Cole-Parmer™ Essentials
0439531 |
100 mL | Non-sterile | 100 |
Case of 100 Each for $201.81
|
|
|||
Description
- Certified RNase-, DNase-, and pyrogen-free
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.
Single Chamber Reservoirs are made of white polystyrene and constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring. Available in 25, 50, and 100 mL volumes, the reservoirs are ideal for 8- and 12-channel pipettors.
The 12-Chamber Reservoir is designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250°F (-80 to 121°C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.
Specifications
| White | |
| Pipetting Reservoir | |
| Not Autoclavable | |
| Single-use |
| Polystyrene | |
| Holding Reagent | |
| 1 | |
| V-bottom |