missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polystyrene for GPC 10,000, MilliporeSigma™ Supelco™
Polystyrene is a synthetic thermoplastic, which can find a wide range of applications due to its properties like low cost, rigidity and good processability.
$393.79 - $393.79
Specifications
| Physical Form | Neat |
|---|---|
| Linear Formula | [CH2 CH(C6H5)]n |
| MDL Number | MFCD00084450 |
| Synonym | PS |
| Type | Analytical Standard |