missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Polystyrene Bulk Wrapped Reagent Reservoir

Catalog No. 03449263
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
55 mL
100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03449263 50 mL
03449256 10 mL
03449260 25 mL
03449279 55 mL
03449267 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03449263 Supplier Heathrow Scientific™ Supplier No. 120645

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

  • Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Quality certificate available on request
  • Disposable

Specifications

Capacity (English) 1.7 oz.
Color White
Capacity (Metric) 50 mL
Dimensions (L x W x H) 4 x 1.1 x 1.2 in.
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir
Packaging Bulk Wrapped
Quantity 100 Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Convoluted
Show More Show Less