missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene, Analytical Standard, For GPC, 900,000, MilliporeSigma™ Supelco™

Catalog No. 111004600
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004600 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004600 Supplier Merck Emd Millipore Supplier No. 3278081G

Polystyrene (for GPC, 900,000) is a synthetic thermoplastic, that is suitable for use in a wide range of applications on account of its characteristics such as low cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.

Specifications

Packaging Glass Bottle
Physical Form Fibers
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Quantity 1 g
Grade GPC