missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene, Analytical Standard, For GPC, 90,000, MilliporeSigma™ Supelco™

Catalog No. 111004660
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004660 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004660 Supplier Merck Emd Millipore Supplier No. 3277781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polystyrene (for GPC, 90,000) is one of the synthetic thermoplastics, used in a broad-range of applications because of its properties like cheap cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.

Specifications

Packaging Glass Bottle
Physical Form Powder
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Quantity 1 g
Grade GPC