missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene, Analytical Standard, For GPC, 200,000, MilliporeSigma™ Supelco™

Catalog No. 111004589
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004589 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004589 Supplier Merck Emd Millipore Supplier No. 3277861G
Only null left

Polystyrene (for GPC, 200,000) is a synthetic thermoplastic, that may be useful for a variety of applications because of its characteristics ike low cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and ease of processability.

Specifications

Packaging Glass Bottle
Physical Form Powder
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Quantity 1 g
Grade GPC