missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 85, Spectrum™ Chemical

$1,875.25 - $3,917.75

Chemical Identifiers

CAS 9005-70-3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612421
View Documents Promotion Details
N/A 4 L Poly Drum
Each for $1,875.25
Only null left
 
18612420
View Documents Promotion Details
N/A 20 L Poly Drum
Each for $3,917.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9005-70-3

Specifications

9005-70-3
39-52
Ungraded
1
80 - 95
Polysorbate 85
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.