missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 85, Spectrum™ Chemical

Catalog No. 18612421
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612421 4 L Poly Drum
18612420 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612421 Supplier Spectrum Chemical Mfg Cor Supplier No. PO1404LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-70-3

Specifications

CAS Max % 1
Quantity 4 L
Hydroxyl Value 39-52
Packaging Poly Drum
Saponification Value 80 - 95
Grade Ungraded
Chemical Name or Material Polysorbate 85

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.