missing translation for 'onlineSavingsMsg'
Learn More

Medisca Polysorbate 80 NF (Tween 80)

Catalog No. 14186045
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186045 500 mL
14185675 1 L
14185563 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186045 Supplier Medisca Supplier No. 052608

Polysorbate 80 NF is a lemon to amber-colored, oily liquid having a faint characteristic odor.

Polysorbate 80 NF is a lemon to amber-colored, oily liquid having a faint characteristic odor.

Specifications

Chemical Name or Material Polysorbate 80 NF (Tween 80)
Packaging Bottle, 500mL, Amber, PET, Boston Round, 28-400
CAS 9005-65-6
Type Drug Standard
Molecular Weight (g/mol) 604.81
Quantity 500 mL
Grade NF