missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Polysorbate 80 NF (Tween 80)
Description
Polysorbate 80 NF is a lemon to amber-colored, oily liquid having a faint characteristic odor.
Specifications
Specifications
| Chemical Name or Material | Polysorbate 80 NF (Tween 80) |
| Packaging | Bottle, 32oz.Amber PET Boston Round, 28-410 |
| CAS | 9005-65-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 604.81 |
| Quantity | 1 L |
| Grade | NF |