missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80, NF, Spectrum™ Chemical
PO138, 9005-65-6
$540.71 - $889.07
Chemical Identifiers
| CAS | 9005-65-6 |
|---|---|
| Molecular Formula | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165986 |
| InChI Key | HMFKFHLTUCJZJO-UHFFFAOYNA-N |
| IUPAC Name | 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Description
Spectrum™ Chemical Polysorbate 80, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 9005-65-6 | |
| 522.68 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| MFCD00165986 | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Specifications
| 9005-65-6 | |
| 1 | |
| 96-108 | |
| 45-55 | |
| 300-500 cSt | |
| MFCD00165986 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Polysorbate 20 |
| 2 | |
| Neutral | |
| 0.0025 | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| 0.03 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| NF |