missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20

Catalog No. p-7046569
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15029AE 100 mL
AAL15029AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15029AE Supplier Thermo Scientific Chemicals Supplier No. L15029AE

CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 20 is used as a wetting agent in flavored mouth drops. It acts as a washing agent in Western blots and ELISAs and prevents antibody binding. It is a solubilizing agent of membrane proteins. In pharmaceutical chemistry, it is used as an excipient to stabilize emulsions and suspensions. Further, it is used as a dispersing agent, stabilizer, lubricator and also used in creams, salves, ointments, protective creams, balms, pomades, lipsticks, mascaras and glosses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
PubChem CID 443314
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Boiling Point >100°C
Density 1.105
Solubility Information Miscible with water.
Quantity 100 mL
Flash Point >110°C (230°F)
Refractive Index 1.47
Chemical Name or Material Polysorbate 20
EINECSNumber 500-018-3
RTECSNumber TR7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only