missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, MP Biomedicals™

Catalog No. ICN19484180
Change view
Click to view available options
Quantity:
100 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19484180 100 mL
ICN19472404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19484180 Supplier MP Biomedicals Inc Supplier No. 0219484180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylenesorbitan monolaurate, Tween 20
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Oily Liquid
Density 1.095 g/mL
Quantity 100 mL
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Chemical Name or Material Polysorbate 20