missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Electrophoresis Reagent

CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol

$55.79 - $83.81

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monolaurate; Tween™ 20
PubChem CID 443314
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ66278AE
View Documents Promotion Details
N/A 100 mL
Each for $55.79
Only null left
 
AAJ66278AP
View Documents Promotion Details
N/A 500 mL
Each for $83.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9005-64-5
522.68
HMFKFHLTUCJZJO-UHFFFAOYNA-N
443314
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MFCD00165986
Polyoxyethylene sorbitan monolaurate; Tween™ 20
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate

Specifications

Colorless to Yellow
Liquid
1.105 g/mL
1.47
MFCD00165986
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
443314
9005-64-5
>100°C
>110°C (230°F)
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Polyoxyethylene sorbitan monolaurate; Tween™ 20
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Polysorbate 20, Electrophoresis Reagent
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 500-018-3

RTECSNumber : TR7400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only