missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Stirring Rods, 12 Pack

Polypropylene stirring rods are ideal for stirring solutions in a variety of vessels within a lab setting.

$24.86 - $47.14

Specifications

Autoclavable Autoclavable
Color White
Material Polypropylene
Type Stirring Rod
For Use With (Application) Stirring
View More Specs

Products 3
Catalog Number Mfr. No. Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Price Quantity  
S41762
View Documents
Eisco™
CH0757B
15 cm
Pack of 12 Each for $24.86
Only null left
 
S41764
View Documents
Eisco™
CH0757C
20 cm
Pack of 12 Each for $37.86
Only null left
 
S41761
View Documents
Eisco™
CH0757A
24.89 cm
Pack of 12 Each for $47.14
Only null left
 
Description

Description

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Autoclavable
Polypropylene
Stirring
White
Stirring Rod
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'