Learn More
Cytiva Polypropylene Screw Lid GL45 Kit
Description
- The kit contains 1 x Screw lid 1 x Cap membrane
- The membrane should not be used when working with organic solvent.
- Lid: Polypropylene (PP)
- Membrane: Ethylenepropylenediene (EPDM)
Specifications
Specifications
| For Use With (Equipment) | Lid |
| Material | Polypropylene, EPDM |
| Includes | Screw lid, Cap membrane |
| Quantity | 1 Kit |