missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Reagent Bottles with Screw Caps

Catalog No. s88423
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
60 mL
125 mL
250 mL
Mouth:
Narrow
Quantity:
1 Ea.
10/Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Mouth Quantity
S88423 4 mL Narrow 10/Pk.
S39295 4 mL Narrow 1 Ea.
S39296 8 mL Narrow 1 Ea.
S39300 60 mL Narrow 1 Ea.
S39302 125 mL Narrow 1 Ea.
S39304 250 mL Narrow 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S88423 Supplier Eisco™ Supplier No. CH0170ANPK10
Only null left

Includes

bottle, threaded screw cap

Transparent, rigid polypropylene reagent bottles with screw caps can be used to store liquid solutions in the laboratory. Threaded screw cap renders the bottles spill and leak proof. Autoclavable. Narrow and wide neck varieties available.

  • Made of high quality, rigid polypropylene (PP)
  • Includes threaded polypropylene screw cap
  • Designed to store liquid solutions in the laboratory
  • Spill and leak resistant
  • Autoclavable
  • Narrow and wide mouth varieties available

Warranty

1 year limited manufacturer warranty

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Clear
Description Dishwasher safe
Diameter (English) 0.6 in.
For Use With (Application) Holding liquids
Includes Screw caps
Height (Metric) With Closure 40.64 mm
Material Polypropylene
Length (Metric) 1.5 cm
Quantity 10/Pk.
Sterility Non-sterile
Width (Metric) 1.5 cm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 4 cm
Product Type Reagent Bottle
Height (English) 1.6 in
Closure Size 0.85 cm
Diameter (Metric) 1.5 cm
Autoclavable Autoclavable
Show More Show Less