missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Polypropylene Reagent Bottles with Screw Caps
Description
Includes
bottle, threaded screw cap
- Made of high quality, rigid polypropylene (PP)
- Includes threaded polypropylene screw cap
- Designed to store liquid solutions in the laboratory
- Spill and leak resistant
- Autoclavable
- Narrow and wide mouth varieties available
Warranty and Services
1 year limited manufacturer warranty
Specifications
Specifications
| Capacity (Metric) | 4 mL |
| Capacity (English) | 0.14 oz. |
| Color | Clear |
| Description | Narrow mouth |
| Diameter (English) | 0.6 in. |
| For Use With (Application) | Solution Storage |
| Includes | Screw caps |
| Height (Metric) With Closure | 40.64 mm |
| Material | Polypropylene |
| Length (Metric) | 15.2 mm |
| Show More |