Learn More
Eisco™ Polypropylene Flat Head Stoppers, Multipack
Acid-proof, autoclavable, and come in a variety of joint sizes
$10.96 - $18.11
Specifications
Autoclavable Autoclavable
Color White
Material Polypropylene
Product Type Plug Stopper
Shape Octagonal Grip
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
S88317
View Documents
Eisco™
CH0873A
10/19
Pack of 10 Each for $10.96
Only null left
S88318
View Documents
Eisco™
CH0873B
12/21
Pack of 10 Each for $14.14
Only null left
S88319
View Documents
Eisco™
CH0873C
14/23
Pack for $16.07
Only null left
S88320
View Documents
Eisco™
CH0873D
19/26
Pack for $18.11
Only null left
S88321
View Documents
Eisco™
CH0873E
24/29
Pack for $17.36
Only null left
S29499
View Documents
Eisco™
CH0873F
29/32
Pack for $17.86
Only null left
Description
Description
  • Lightweight and durable polypropylene stopper
  • Chemical resistant, acid proof
  • Flat head design
  • Used to seal glass flasks and bottles
  • Autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
Autoclavable
Polypropylene
Octagonal Grip
Seal flasks
White
Plug Stopper
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'