missing translation for 'onlineSavingsMsg'
Learn More

Polyoxyethylene bis(amine), M.W. 1,000

Catalog No. p-7080218
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46833MD 250 mg
AA4683303 1 g
AA4683306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46833MD Supplier Thermo Scientific Chemicals Supplier No. 046833MD
Only null left

CAS: 24991-53-5 | H2NCH2CH2(OCH2CH2)nNH2

It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24991-53-5
Molecular Formula H2NCH2CH2(OCH2CH2)nNH2
MDL Number MFCD00132004
Synonym PEG-diamine

Specifications

Color White
Physical Form Granular Solid
Solubility Information It is soluble in organic solvents. It is insoluble in water.
Quantity 250 mg
Sensitivity Air Sensitive
Chemical Name or Material Polyoxyethylene bis (amine), M.W. 1,000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only