accessibility menu, dialog, popup

UserName

H2NOONH2

CAS RN 24991-53-5

IUPAC Name: 2-[2-(2-aminoethoxy)ethoxy]ethan-1-amine
Synonyms: 2-[2-(2-aminoethoxy)ethoxy]ethanamine
Molecular Weight (g/mol): 148.21
Molecular Formula: C6H16N2O2
InChi Key: IWBOPFCKHIJFMS-UHFFFAOYSA-N
SMILES: NCCOCCOCCN
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)