missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polynuclear aromatic hydrocarbons 16 solution, in cycloHexane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 110-82-7 |
| MDL Number | MFCD00148504 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Grade | Analytical Standard |