missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013054
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013054 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013054 Supplier Merck Emd Millipore Supplier No. PHR1595500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polymyxin B Sulfate is used for the treatment of gram-negative bacterial infections caused particularly by Pseudomonas aeruginosa and Escherichia coli.

Specifications

CAS 1405-20-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Synonym Polymyxin B sulfate
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material