missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polymyxin B Sulfate MP Biomedicals
$1,177.22
Chemical Identifiers
| CAS | 1405-20-5 |
|---|---|
| Molecular Formula | C48H84N16O17S |
| Molecular Weight (g/mol) | 1189.36 |
| MDL Number | MFCD00131991 |
| InChI Key | HNDFYNOVSOOGDU-UHFFFAOYNA-N |
| Synonym | Aerosporin |
| PubChem CID | 133109994 |
| IUPAC Name | N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox |
| SMILES | OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10056550
|
N/A | 50 mU |
Each for $1,177.22
|
|
|||||
Chemical Identifiers
| 1405-20-5 | |
| 1189.36 | |
| HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
| 133109994 | |
| OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
| C48H84N16O17S | |
| MFCD00131991 | |
| Aerosporin | |
| N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox |
Specifications
| Polymixin B Sulfate | |
| 1405-20-5 | |
| C48H84N16O17S | |
| Aerosporin | |
| OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O | |
| 1189.36 | |
| 1385.61 |
| Powder | |
| White | |
| MFCD00131991 | |
| HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
| N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox | |
| 133109994 | |
| Polymyxin B Sulfate, Aerosporin Activity: >6, 000 u/mg |
Safety and Handling
Recommended Storage : Store at 2° to 8°C.