missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Solution, 1 mg/mL in Water, MilliporeSigma™ Supelco™

Catalog No. 111013053
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013053 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013053 Supplier Merck Emd Millipore Supplier No. 8127110ML

Polymyxin B solution can find therapeutic applications as an antimicrobial agent, since it shows activity against multidrug-resistant (MDR) gram-negative bacterial infections.

Specifications

CAS 1405-20-5
Concentration 1 mg/mL in H 2 O
MDL Number MFCD00131991
Type Analytical Standard
Quantity 10 mL
Grade Analytical Standard