Learn More
Polymedco™ Sediplast™ Autozero ESR System
Closed system with unique vented cap: precisely calibrated hole stops the column of blood at true zero
Supplier:  Polymedco™ S 1045 2
Encompass
 View more versions of this product
Catalog No. 0267027
Only null left
Description
Description
  • Pierceable stopper manufactured from a special formulation of linear low-density polyethylene to ensure a leakproof seal during the procedure
  • When the pipet is inserted into the vial, the diaphragm is designed to break away and the unit becomes a completely sealed system
  • Eliminates all manipulation “techniques” in setting sample at the zero mark
  • All excess blood flows into a closed, separate reservoir compartment automatically
  • Includes a stoppered, pre-filled vial containing 0.2mL of 3.8% sodium citrate diluent
  • Simply add 0.8mL of blood and mix
  • Eliminates separate diluent preparation, dilution, and handling steps required in other systems
  • Also available without diluent
Specifications
Chemical Identifiers
Whole EDTA Blood
Used for hematology applications
SEDIPLAST Autozero ESR System
Specifications
Acrylic Rack
Whole EDTA Blood
Used for hematology applications
SEDIPLAST Autozero ESR System
Product Suggestions
Product Suggestions