missing translation for 'onlineSavingsMsg'
Learn More

Polyimide film, 0.013mm thick, Thermo Scientific Chemicals

Catalog No. AA41897GH
Change view
Click to view available options
Quantity:
100 x 100 mm
150 x 150 mm
300 x 300 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41897GH 100 x 100 mm
AA41897GT 150 x 150 mm
AA41897HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41897GH Supplier Thermo Scientific Chemicals Supplier No. 41897GH

Polyimide film is used in electronic and integrated circuit industries. It is also used for the reverse osmotic film in purification of water and the concentration of dilute materials from water. It finds application in making high temperature tape, IC Passivation coating, LCD alignment layer and insulated materials. It plays an important role in consumer electronic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62929-02-6
Molecular Formula C35H28N2O7
Molecular Weight (g/mol) 588.62
MDL Number MFCD00677744
InChI Key CDTDIQLZIBORMV-UHFFFAOYNA-N
Synonym polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione
PubChem CID 6454485
SMILES CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Physical Form ≈0.1846g/100x100mm
Solubility Information Insoluble in water.
Quantity 100 x 100 mm
IUPAC Name 1-(4-aminophenyl)-1,3,3-trimethyl-2,3-dihydro-1H-inden-5-amine; 5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl)-1,3-dihydro-2-benzofuran-1,3-dione
Chemical Name or Material Polyimide film, 0.013mm thick

RUO – Research Use Only