missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041896GT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Polyimide film | |
| ≈0.1136g/100x100mm | |
| C35H28N2O7 | |
| polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione | |
| CDTDIQLZIBORMV-UHFFFAOYNA-N | |
| 1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione | |
| 6454485 |
| 0.008mm thick | |
| 62929-02-6 | |
| MFCD00677744 | |
| 150 x 150 mm | |
| CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1 | |
| 588.62 |
Chemical Identifiers
| 62929-02-6 | |
| 588.62 | |
| CDTDIQLZIBORMV-UHFFFAOYNA-N | |
| 6454485 | |
| CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1 |
| C35H28N2O7 | |
| MFCD00677744 | |
| polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione | |
| 1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione |
Safety and Handling
TSCA : Yes
RUO – Research Use Only