Learn More
Bel-Art™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier:  Bel-Art™ H131781016
 View more versions of this product
Catalog No. 13-717-282
Only null left
Explore available promotions
Description
Description
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
Not Autoclavable
Clear
Polyethylene
0.102 mm
10 in.
16 in.
Sample Bag
Zip Tie
Single-use
Non-sterile
10 x 16 in. (254 x 406mm)
254 mm
406 mm
Product Suggestions
Product Suggestions