missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyethylene Safety Bottle Carriers

Safe, simple way to carry bottles.

 View more versions of this product

Catalog No. 0343975B


Only null left

Description

Description

Safe, simple way to carry bottles.

  • Safe, simple way to carry bottles
  • Made of heavy-duty HDPE
  • One inch diameter carrying handle

Order Info

Dimensions, 14.5L x 7.5W x 6.25 in. H (36.8 x 19.1 x 15.9cm). Additional bottle carrier sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Chemical Identifiers

Not Autoclavable
HDPE
36.8 cm
7.5 in.
Bottle Carrier
White
No
14.5 in.
6.25 in.
19.1 cm

Specifications

Not Autoclavable
Bottle (100 mL)
6
14.5 x 7.5 x 6.25 in. (36.8 x 19.1 x 15.24 cm)
14.5 in.
6.25 in.
15.24 cm
White
HDPE
No
36.8 cm
7.5 in.
Bottle Carrier
19.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions