missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 1,000,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Supplier:  Thermo Scientific Chemicals 04367822

 View more versions of this product

Catalog No. AA4367822


Only null left
Explore available promotions

Description

Description

Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polyethylene oxide
5 to 7
25322-68-3
MFCD01779601
Soluble in organic solvents,aromatic hydrocarbons and water. Slightly soluble in aliphatic hydrocarbons.
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
Odorless
M.W. 1,000,000
Powder
(C2H4O)n
14,7568
100 g
[H]OCCO
62.07

Chemical Identifiers

25322-68-3
LYCAIKOWRPUZTN-UHFFFAOYSA-N
[H]OCCO
62.07
ethane-1,2-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 500-038-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only