missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 1,000,000

Catalog No. p-7058963
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4367814 25 g
AA4367822 100 g
AA4367836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4367814 Supplier Thermo Scientific Chemicals Supplier No. 04367814
Only null left

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Physical Form Powder
Molecular Formula (C2H4O)n
MDL Number MFCD01779601
Merck Index 14,7568
Solubility Information Soluble in organic solvents,aromatic hydrocarbons and water. Slightly soluble in aliphatic hydrocarbons.
Quantity 25 g
Odor Odorless
Chemical Name or Material Polyethylene oxide, M.W. 1,000,000
EINECSNumber 500-038-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only