missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene, low density, ∣<400 micron

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

Supplier:  Thermo Scientific Chemicals 0426070E

 Afficher plus de versions de ce produit

Numéro de catalogue. AA426070E


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Polyethylene is used in injection and blow molded toys, housewares, lids, injection molded car seats, Hot melt coating for paper, additive in cast moldings, candles, oil-based inks and hot melt adhesives. It is also used as an additive, lubricant in rubber processing, extrusion and calendaring aid for PVC and dispersing aid for color concentrates. Bearings, gears, bushings and other moving parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Polyethylene
Powder
(C2H4)n
0.92 g/mL
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
2500 g
*-CC-*
28.05
CHEBI:18153
Low density, ≤400 micron
9002-88-4
MFCD00084423
14,7567
Insoluble in water.
VGGSQFUCUMXWEO-UHFFFAOYSA-N
ethene
6325
Odorless

Identifiants chimiques

9002-88-4
28.05
VGGSQFUCUMXWEO-UHFFFAOYSA-N
6325
ethene
(C2H4)n
MFCD00084423
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
CHEBI:18153
*-CC-*
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'rtecsNumber' : TQ3325000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only