missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene, low density, 1000 micron

Catalog No. p-7059097
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4394918 50 g
AA4394930 250 g
AA43949A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4394918 Supplier Thermo Scientific Chemicals Supplier No. 04394918
Only null left

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

Polyethylene is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for the manufacturing of various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipments. It is used in the manufacture of plastic bags, yard bags or outdoor bags, which is an environmentally sensible material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-88-4
Molecular Formula (C2H4)n
Molecular Weight (g/mol) 28.05
MDL Number MFCD00084423
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
PubChem CID 6325
ChEBI CHEBI:18153
IUPAC Name ethene
SMILES *-CC-*

Specifications

Physical Form Granular
Merck Index 14,7567
Solubility Information Insoluble in water.
Quantity 50 g
Odor Odorless
Chemical Name or Material Polyethylene, Low density, 1000 micron
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only