missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol/Polyethylene oxide ReadyCal Set Mp ∼250 -1,000,000, MilliporeSigma™ Supelco™

Catalog No. 111013736
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013736 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013736 Supplier Merck Emd Millipore Supplier No. 023931EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PEG also known as ethylene glycol, is a colorless, odorless, viscous dihydroxy alcohol. It is a polyether compound with applications ranging from industrial manufacturing to medicine. It is the most important glycol that is commercially available, and finds application as an antifreeze and coolant, in hydraulic fluids, and in the manufacture of low-freezing dynamites and resins. It is widely used as covalent modifier of biological macromolecules.

Specifications

CAS 25322-68-3
Linear Formula H(OCH2 CH2)nOH
MDL Number MFCD00081839
Synonym Poly(ethylene glycol); PEG
Type Analytical Standard
Quantity 1 Ea.
Grade Analytical Standard