missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monododecyl Ether (n=approx. 25), TCI America™

Catalog No. P067125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P067125G 25 g
P0671500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P067125G Supplier TCI America Supplier No. P067125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyethylene Glycol Monolauryl Ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color White-Yellow
Physical Form Crystal at 20°C
Quantity 25 g
Chemical Name or Material Polyethylene Glycol Monododecyl Ether (n=approx. 25)
EINECSNumber (7)-0097
RTECSNumber JR5990000
TSCA Yes