missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 8000, NF, Spectrum™ Chemical
PO131, 25322-68-3, H(OCH2CH2)nOH
$917.77 - $6657.33
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605346
|
Spectrum Chemical Mfg Cor
PO13125KG |
2.5 kg | Poly Pail |
Each for $917.77
|
|
||||
|
18605347
|
Spectrum Chemical Mfg Cor
PO13112KG |
12 kg | Poly Pail |
Each for $3,152.20
|
|
||||
|
18605348
|
Spectrum Chemical Mfg Cor
PO13145KGBLPL |
45 kg | Poly Drum |
Each for $6,657.33
|
|
||||
Description
Spectrum™ Chemical Polyethylene Glycol 8000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| [H]OCCO |
| (C2H4O)n | |
| MFCD01779601 | |
| ethane-1,2-diol |
Specifications
| 4.5 to 7.5 | |
| 1 | |
| MFCD01779601 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| NF | |
| Poly Pail |
| 25322-68-3 | |
| (C2H4O)n | |
| 2.5 kg | |
| [H]OCCO | |
| 62.07 | |
| 0.001 | |
| 470-900 cSt |