missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 8000, NF, Spectrum™ Chemical
PO131, 25322-68-3, H(OCH2CH2)nOH
Supplier: Spectrum Chemical Mfg Cor PO13112KG
Description
Spectrum™ Chemical Polyethylene Glycol 8000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 4.5 to 7.5 | |
| 1 | |
| MFCD01779601 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| NF | |
| Poly Pail |
| 25322-68-3 | |
| (C2H4O)n | |
| 12 kg | |
| [H]OCCO | |
| 62.07 | |
| 0.001 | |
| 470-900 cSt |
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| [H]OCCO |
| (C2H4O)n | |
| MFCD01779601 | |
| ethane-1,2-diol |