missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 400, NF, Spectrum™ Chemical
PO110, 25322-68-3, H(OCH2CH2)nOH
$200.97 - $4,420.53
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605336
|
Spectrum Chemical Mfg Cor
PO11025ML |
25 mL | Amber Glass Bottle |
N/A
|
|
||||
|
18605333
|
Spectrum Chemical Mfg Cor
PO11025LTGL |
2.5 L | Glass Bottle |
N/A
|
|
||||
|
18605334
|
Spectrum Chemical Mfg Cor
PO11020LT |
20 L | Poly Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Polyethylene Glycol 400, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| [H]OCCO |
| (C2H4O)n | |
| MFCD01779601 | |
| ethane-1,2-diol |
Specifications
| 4.5 to 7.5 | |
| 1 | |
| MFCD01779601 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| NF | |
| Amber Glass Bottle |
| 25322-68-3 | |
| (C2H4O)n | |
| 25 mL | |
| [H]OCCO | |
| 62.07 | |
| 0.001 | |
| 6.8-8.0 cSt |