missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 400, NF, Spectrum™ Chemical

Catalog No. 18605336
Change view
Click to view available options
Quantity:
25 mL
20 L
Packaging:
Amber Glass Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605336 25 mL Amber Glass Bottle
18605334 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605336 Supplier Spectrum Chemical Mfg Cor Supplier No. PO11025ML
Only null left

PO110, 25322-68-3, H(OCH2CH2)nOH

Spectrum™ Chemical Polyethylene Glycol 400, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 4.5 to 7.5
CAS Max % 1
Quantity 25 mL
Grade NF
Ignition Residue 0.001
Packaging Amber Glass Bottle
Viscosity 6.8-8.0 cSt