missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 300, TCI America™
$35.41 - $55.92
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | PEG 300 |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG 300 | |
| [H]OCCO |
Specifications
| Colorless | |
| 25322-68-3 | |
| MFCD01779601 | |
| 25 g | |
| [H]OCCO | |
| 62.07 |
| Clear Liquid at 20°C | |
| (C2H4O)n | |
| PEG 300 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| Polyethylene Glycol 300 |
Safety and Handling
EINECSNumber : (7)-0129
RTECSNumber : TQ3630000
TSCA : Yes