missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 300, TCI America™

Supplier:  TCI America H054325G

 View more versions of this product

Catalog No. H054325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Colorless
Clear Liquid at 20°C
(C2H4O)n
PEG 300
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
Polyethylene Glycol 300
25322-68-3
MFCD01779601
25 g
[H]OCCO
62.07

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
(C2H4O)n
MFCD01779601
PEG 300
[H]OCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0129

RTECSNumber : TQ3630000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions