missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 1450, NF, Spectrum™ Chemical
P1492, 25322-68-3, H(OCH2CH2)nOH
$298.00 - $6029.70
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605303
|
Spectrum Chemical Mfg Cor
P1492500GM |
500 g | Poly Bottle |
Each for $298.00
|
|
||||
|
18605304
|
Spectrum Chemical Mfg Cor
P149225KG |
2.5 kg | Poly Pail |
Each for $823.93
|
|
||||
|
18605305
|
Spectrum Chemical Mfg Cor
P149212KGBL |
12 kg | Poly Pail |
Each for $3,001.17
|
|
||||
|
18605306
|
Spectrum Chemical Mfg Cor
P149245KGBL |
45 kg | Fiber Drum |
Each for $6,029.70
|
|
||||
Description
Spectrum™ Chemical Polyethylene Glycol 1450, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| [H]OCCO |
| (C2H4O)n | |
| MFCD01779601 | |
| ethane-1,2-diol |
Specifications
| 4.5 to 7.5 | |
| 1 | |
| MFCD01779601 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| NF | |
| Poly Bottle |
| 25322-68-3 | |
| (C2H4O)n | |
| 500 g | |
| [H]OCCO | |
| 62.07 | |
| 0.001 | |
| 25-32 cSt |