missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 1450, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Polyethylene Glycol 1450, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| pH | 4.5 to 7.5 |
| CAS Max % | 1 |
| Quantity | 45 kg |
| Grade | NF |
| Ignition Residue | 0.001 |
| Packaging | Fiber Drum |
| Viscosity | 25-32 cSt |