missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Polyethylene Frit, Porosity: 20 μm
Frits are crucial components of the high performance liquid chromatography system design. They act as a depth filter for the concurrent physical removal of small molecules. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.
Supplier: MilliporeSigma™ Supelco™ 57183
Chemical Identifiers
| Crystalline Precipitates, Extraction | |
| Polyethylene | |
| 20 μm | |
| Frit |
Specifications
| Crystalline Precipitates, Extraction | |
| Polyethylene | |
| 20 μm |
| 20 mL SPE Tube | |
| 40 | |
| Frit |