Learn More
MilliporeSigma™ Supelco™ Polyethylene Frit, Porosity: 20 μm
Frits are crucial components of the high performance liquid chromatography system design. They act as a depth filter for the concurrent physical removal of small molecules. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.
Supplier:  MilliporeSigma™ Supelco™ 57181
 View more versions of this product
Catalog No. 111004909
Only null left
Description
Description
Specifications
Chemical Identifiers
Crystalline Precipitates, Extraction
Polyethylene
20 μm
Frit
Specifications
Crystalline Precipitates, Extraction
Polyethylene
20 μm
6 mL SPE Tube
60
Frit
Product Suggestions
Product Suggestions