missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyethylene Cleanroom Clipboards

Catalog No. 18003067
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
Teal
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18003067 Black
18003069 Blue
18003071 Green
18003072 Orange
18003068 Purple
18003073 Red
18003074 Teal
18003070 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18003067 Supplier Fisherbrand™ Supplier No. CLBOARD11BLA

Polyethylene cleanroom clipboards with poly clip. Available in 9 colors, for 8.5 x 11 in. documents.

  • Durable and minimal
  • Pre-wiped down with IPA
  • ESD-safe

Specifications

Material Polyethylene
Color Black
For Use With (Equipment) 8.5 x 11 in. documents
Product Type Cleanroom Clipboard