missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Adhesive Warning Sign: HAZARDOUS AREA AUTHORIZED PERSONNEL ONLY

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

Supplier:  Brady™ 124685

 View more versions of this product

Catalog No. 19-105-024

Only null left
Explore available promotions

Description

Description

  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications

Specifications

Laminated
1 Ea.
Self-adhesive
230°F
0.01 in.
Rectangular
10 x 7 in.
Black on Yellow
Polyester
Cold-temperature Permanent Acrylic
7 in.
-40°F
10 in.
-40°F to 230°F
HAZARDOUS AREA AUTHORIZED PERSONNEL ONLY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions